Please login to your online account to display your discounted pricing

Bis(2-methoxyethyl) Ether (Certified), Fisher Chemical


Quantity 4L

Manufacturer: fisher scientific company  O14714

 View more versions of this product

Catalog No. O1471-4

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99%
Melting Point −64°C
Purity Grade Certified Grade
Formula Weight 134.18
Boiling Point 162°C
Quantity 4L
DOT Information DOT Class 3, : Flammable Liquid
Density 0.937g/cm³
Physical Form Liquid
Vapor Density 4.62g/mol
Color Clear
Molecular Formula C6H14O3
UN Number 3271
Synonym Diethylene Glycol Dimethyl Ether
Boiling Range 2°C max.
Identification Pass Test
Linear Formula CH3OCH2CH2OCH2CH2OCH3
CAS 111-96-6
Chemical Name or Material Bis(2-methoxyethyl) Ether
MDL Number MFCD00008503
Packaging Glass Bottle
Assay Percent Range ≥99 %
ChemAlert Storage Symbol Red

Specifications

Chemical Name or Material Bis(2-methoxyethyl) Ether
CAS 111-96-6
Purity Grade Certified Grade
Assay ≥99%
Assay Percent Range ≥99 %
Molecular Formula C6H14O3
Linear Formula CH3OCH2CH2OCH2CH2OCH3
Formula Weight 134.18
MDL Number MFCD00008503
Synonym Diethylene Glycol Dimethyl Ether
Accessories
 

SDS & Safety Documents