missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bis(2-methoxyethyl) Ether (Certified), Fisher Chemical™
$353.23 - $3,251.54

Chemical Identifiers
CAS | 111-96-6 |
---|---|
Molecular Formula | C6H14O3 |
Molecular Weight (g/mol) | 134.175 |
MDL Number | MFCD00008503 |
InChI Key | SBZXBUIDTXKZTM-UHFFFAOYSA-N |
Synonym | 1-methoxy-2-2-methoxyethoxy ethane, 2,5,8-trioxanonane, 2-methoxyethyl ether, bis 2-methoxyethyl ether, di 2-methoxyethyl ether, diethylene glycol dimethyl ether, diglyme, dimethyl carbitol, poly-solv |
PubChem CID | 8150 |
ChEBI | CHEBI:46784 |
IUPAC Name | 1-methoxy-2-(2-methoxyethoxy)ethane |
SMILES | COCCOCCOC |
Chemical Identifiers
111-96-6 | |
134.175 | |
SBZXBUIDTXKZTM-UHFFFAOYSA-N | |
8150 | |
1-methoxy-2-(2-methoxyethoxy)ethane |
C6H14O3 | |
MFCD00008503 | |
1-methoxy-2-2-methoxyethoxy ethane, 2,5,8-trioxanonane, 2-methoxyethyl ether, bis 2-methoxyethyl ether, di 2-methoxyethyl ether, diethylene glycol dimethyl ether, diglyme, dimethyl carbitol, poly-solv | |
CHEBI:46784 | |
COCCOCCOC |
Specifications
2°C max. | |
Pass Test | |
162°C | |
-64°C | |
111-96-6 | |
C6H14O3 | |
MFCD00008503 | |
1-methoxy-2-2-methoxyethoxy ethane, 2,5,8-trioxanonane, 2-methoxyethyl ether, bis 2-methoxyethyl ether, di 2-methoxyethyl ether, diethylene glycol dimethyl ether, diglyme, dimethyl carbitol, poly-solv | |
COCCOCCOC | |
134.175 | |
CHEBI:46784 | |
Liquid | |
Certified |
0.937g/cm³ | |
Glass Bottle | |
Undesignated | |
500mL | |
≥99 % | |
CH3OCH2CH2OCH2CH2OCH3 | |
3271 | |
SBZXBUIDTXKZTM-UHFFFAOYSA-N | |
1-methoxy-2-(2-methoxyethoxy)ethane | |
8150 | |
134.18 | |
≥99% | |
Bis(2-methoxyethyl) Ether |
Safety and Handling

DOTInformation : DOT Class 3, : Flammable Liquid