missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-methoxyethyl) ether, 99+%, Extra Dry, AcroSeal™

CAS: 111-96-6 | C6H14O3 | 134.18 g/mol

Supplier:  Thermo Scientific Chemicals 397210010

 View more versions of this product

Catalog No. AC397210010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Bis(2-methoxyethyl) ether
111-96-6
100.0
Colorless
162.0°C
Authentic
AcroSeal™ Glass Bottle
1.4070 to 1.4090
MFCD00008503
01, IV, 2393
01,255
15, 3184
Solubility in water: miscible. Other solubilities: miscible with alcohol,ether,hydrocarbon solvents
COCCOCCOC
134.18
2 mPa.s (20°C)
134.18
Extra Dry
Extra Dry
99.0
-64.0°C
0.9370g/mL
51°C
99% min. (GC)
C6H14O3
(CH3OCH2CH2)2O
1 L
0.001 max.
0.937
diglyme, bis 2-methoxyethyl ether, 2-methoxyethyl ether, 1-methoxy-2-2-methoxyethoxy ethane, diethylene glycol dimethyl ether, poly-solv, di 2-methoxyethyl ether, dimethyl carbitol, 2,5,8-trioxanonane
SBZXBUIDTXKZTM-UHFFFAOYSA-N
1-methoxy-2-(2-methoxyethoxy)ethane
8150
CHEBI:46784
99+%
Liquid

Chemical Identifiers

111-96-6
134.18
SBZXBUIDTXKZTM-UHFFFAOYSA-N
8150
1-methoxy-2-(2-methoxyethoxy)ethane
C6H14O3
MFCD00008503
diglyme, bis 2-methoxyethyl ether, 2-methoxyethyl ether, 1-methoxy-2-2-methoxyethoxy ethane, diethylene glycol dimethyl ether, poly-solv, di 2-methoxyethyl ether, dimethyl carbitol, 2,5,8-trioxanonane
CHEBI:46784
COCCOCCOC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May damage fertility or the unborn child.
Flammable liquid and vapor.
May form explosive peroxides.

GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Use personal protective equipment as required.

GHS Signal Word: Danger

EINECSNumber : 203-924-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only