Please login to your online account to display your discounted pricing

[Bis(2-methoxyethyl)amino]sulfur trifluoride, 2.7M (50 wt.%) solution in toluene, ACROS Organics™


Quantity 25g
Packaging Plastic Bottle in Tin Can

Manufacturer: thermo fisher scientific  AC368960250

 View more versions of this product

Catalog No. AC368960250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥95% (NMR, neat substance)
Physical Form Liquid
Formula Weight 221.24
Quantity 25g
Color Dark yellow-orange
Packaging Plastic Bottle in Tin Can
Density 1.2g/mL
CAS Min % 49.0
CAS Max % 51.0
CAS Min % 49.0
CAS Max % 51.0
Linear Formula (CH3OCHCH)nSF3
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
May be fatal if swallowed and enters airways.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
May cause damage to organs through prolonged or repeated exposure.
Suspected of damaging the unborn child.
Highly
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Store in a dry place. Store in a closed container.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C6H14F3NO2S
Chemical Name or Material [Bis(2-methoxyethyl)amino]sulfur trifluoride
CAS 202289-38-1
CAS 108-88-3
Synonym Deoxo-Fluor«
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material [Bis(2-methoxyethyl)amino]sulfur trifluoride
CAS 202289-38-1
CAS Min % 49.0
CAS Max % 51.0
CAS 108-88-3
CAS Min % 49.0
CAS Max % 51.0
Assay ≥95% (NMR, neat substance)
Molecular Formula C6H14F3NO2S
Linear Formula (CH3OCHCH)nSF3
Formula Weight 221.24
Synonym Deoxo-Fluor«
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Store in a dry place. Store in a closed container.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
May be fatal if swallowed and enters airways.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
May cause damage to organs through prolonged or repeated exposure.
Suspected of damaging the unborn child.
Highly

SDS & Safety Documents