missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(2-methoxyethyl) adipate, 98%

$55.51 - $178.89

Chemical Identifiers

CAS 106-00-3
Molecular Formula C12H22O6
Molecular Weight (g/mol) 262.302
MDL Number MFCD00041905
InChI Key GVRNUDCCYWKHMV-UHFFFAOYSA-N
Synonym bis 2-methoxyethyl adipate, hexanedioic acid, bis 2-methoxyethyl ester, bis 2-methoxyethyl hexanedioate, di 2-methoxyethyl adipate, adipic acid, bis 2-methoxyethyl ester, adipic acid bis 2-methoxyethyl ester, bis 2-methoxyethyl adipate, hexanedioic acid, 1,6-bis 2-methoxyethyl ester, 1,6-bis 2-methoxyethyl hexanedioate, di 2-methoxyethyl adipate
PubChem CID 66046
IUPAC Name bis(2-methoxyethyl) hexanedioate
SMILES COCCOC(=O)CCCCC(=O)OCCOC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1049714 View Documents Alfa Aesar™
A1049714
25g Each for $55.51
 
AAA1049722 View Documents Alfa Aesar™
A1049722
100g Each for $178.89
 
Specifications

Chemical Identifiers

106-00-3
262.302
GVRNUDCCYWKHMV-UHFFFAOYSA-N
66046
COCCOC(=O)CCCCC(=O)OCCOC
C12H22O6
MFCD00041905
bis 2-methoxyethyl adipate, hexanedioic acid, bis 2-methoxyethyl ester, bis 2-methoxyethyl hexanedioate, di 2-methoxyethyl adipate, adipic acid, bis 2-methoxyethyl ester, adipic acid bis 2-methoxyethyl ester, bis 2-methoxyethyl adipate, hexanedioic acid, 1,6-bis 2-methoxyethyl ester, 1,6-bis 2-methoxyethyl hexanedioate, di 2-methoxyethyl adipate
bis(2-methoxyethyl) hexanedioate

Specifications

190°C (11mmHg)
106-00-3
(CH2)4(CO2CH2CH2OCH3)2
bis 2-methoxyethyl adipate, hexanedioic acid, bis 2-methoxyethyl ester, bis 2-methoxyethyl hexanedioate, di 2-methoxyethyl adipate, adipic acid, bis 2-methoxyethyl ester, adipic acid bis 2-methoxyethyl ester, bis 2-methoxyethyl adipate, hexanedioic acid, 1,6-bis 2-methoxyethyl ester, 1,6-bis 2-methoxyethyl hexanedioate, di 2-methoxyethyl adipate
COCCOC(=O)CCCCC(=O)OCCOC
262.302
262.3
Bis (2-methoxyethyl) adipate
25g
C12H22O6
MFCD00041905
GVRNUDCCYWKHMV-UHFFFAOYSA-N
bis(2-methoxyethyl) hexanedioate
66046
98%
Safety and Handling

Safety and Handling

TSCA : TSCA