missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2-hydroxyethyl) Terephthalate 85.0+%, TCI America™
Supplier: TCI America B3429500G
Specifications
| Bis(2-hydroxyethyl) Terephthalate | |
| 106°C | |
| C12H14O6 | |
| 500 g | |
| QPKOBORKPHRBPS-UHFFFAOYSA-N | |
| bis(2-hydroxyethyl) benzene-1,4-dicarboxylate | |
| 13739 | |
| ≥85.0% (HPLC) |
| 959-26-2 | |
| White | |
| MFCD00081066 | |
| Terephthalic Acid Bis(2-hydroxyethyl) Ester, BHET | |
| C1=CC(=CC=C1C(=O)OCCO)C(=O)OCCO | |
| 254.238 | |
| 254.24 | |
| Crystalline Powder |
Chemical Identifiers
| 959-26-2 | |
| 254.238 | |
| QPKOBORKPHRBPS-UHFFFAOYSA-N | |
| 13739 | |
| C1=CC(=CC=C1C(=O)OCCO)C(=O)OCCO |
| C12H14O6 | |
| MFCD00081066 | |
| Terephthalic Acid Bis(2-hydroxyethyl) Ester, BHET | |
| bis(2-hydroxyethyl) benzene-1,4-dicarboxylate |
Safety and Handling
EINECSNumber : (3)-1322&(3)-1327
TSCA : Yes