Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis[2-hydroxy-5-methyl-3-(1-methylcyclohexyl)phenyl]methane, 94%

$33.15 - $274.23

Chemical Identifiers

Chemical Name or Material Bis[2-hydroxy-5-methyl-3-(1-methylcyclohexyl)phenyl]methane
CAS 77-62-3
Molecular Formula C29H40O2
Formula Weight 420.64
MDL Number MFCD00151797
Beilstein 2313064
Synonym 2,2'-Methylenebis[6-(1-methylcyclohexyl)-p-cresol]
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1332318

  SDS
alfa aesar™
L1332318
50g Each for $33.15

AAL133230B

  SDS
alfa aesar™
L133230B
1000g Each for $274.23

AAL1332330

  SDS
alfa aesar™
L1332330
250g Each for $98.55
Description & Specifications

Specifications

Formula Weight 420.64
Assay 94%
Molecular Formula C29H40O2
MDL Number MFCD00151797
CAS 77-62-3
Synonym 2,2'-Methylenebis[6-(1-methylcyclohexyl)-p-cresol]
Melting Point 130° to 135°C
Quantity 50g
Odor Odorless
RTECS Number SM0400000
Chemical Name or Material Bis[2-hydroxy-5-methyl-3-(1-methylcyclohexyl)phenyl]methane
TSCA TSCA
Beilstein 2313064