missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis[2-hydroxy-5-methyl-3-(1-methylcyclohexyl)phenyl]methane, 94%, Alfa Aesar™

Manufacturer:  Alfa Aesar L133230B

 View more versions of this product

Catalog No. AAL133230B



Specifications

Specifications

Bis[2-hydroxy-5-methyl-3-(1-methylcyclohexyl)phenyl]methane
130°C to 135°C
77-62-3
MFCD00151797
2,2'-methylenebis 6-1-methylcyclohexyl-p-cresol, ionox wsp, nonox wsp, bisalkofen mtsp, unii-7tir91vdi9, 7tir91vdi9, 2,2'-methylenebis 6-1-methylcyclohexyl-p-cresol, p-cresol, 2,2'-methylenebis 6-1-methylcyclohexyl, 2,2'-methylenebis 4-methyl-6-1-methylcyclohexyl phenol, 2,2'-methylenebis 4-methyl-6-1-methylcyclohexyl phenol
CC1=CC(=C(C(=C1)C2(CCCCC2)C)O)CC3=C(C(=CC(=C3)C)C4(CCCCC4)C)O
420.637
420.64
Odorless
1000g
C29H40O2
2313064
PHXLONCQBNATSL-UHFFFAOYSA-N
2-[[2-hydroxy-5-methyl-3-(1-methylcyclohexyl)phenyl]methyl]-4-methyl-6-(1-methylcyclohexyl)phenol
6486
94%

Chemical Identifiers

77-62-3
420.637
PHXLONCQBNATSL-UHFFFAOYSA-N
6486
CC1=CC(=C(C(=C1)C2(CCCCC2)C)O)CC3=C(C(=CC(=C3)C)C4(CCCCC4)C)O
C29H40O2
MFCD00151797
2,2'-methylenebis 6-1-methylcyclohexyl-p-cresol, ionox wsp, nonox wsp, bisalkofen mtsp, unii-7tir91vdi9, 7tir91vdi9, 2,2'-methylenebis 6-1-methylcyclohexyl-p-cresol, p-cresol, 2,2'-methylenebis 6-1-methylcyclohexyl, 2,2'-methylenebis 4-methyl-6-1-methylcyclohexyl phenol, 2,2'-methylenebis 4-methyl-6-1-methylcyclohexyl phenol
2-[[2-hydroxy-5-methyl-3-(1-methylcyclohexyl)phenyl]methyl]-4-methyl-6-(1-methylcyclohexyl)phenol
Safety and Handling

Safety and Handling

RTECSNumber : SM0400000

TSCA : TSCA

Documents