Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(2-ethylhexyl) sulfoxide

$113.16 - $113.16

Chemical Identifiers

Chemical Name or Material Bis(2-ethylhexyl) sulfoxide
CAS 82374-34-3
Molecular Formula C16H34OS
Formula Weight 274.51g/mol
MDL Number MFCD00040086
Beilstein 4973978
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1129306

 
alfa aesar™
L11293-06
5g Each for $113.16
Description & Specifications

Specifications

Quantity 5g
Quantity 5g
Chemical Name or Material Bis(2-ethylhexyl) sulfoxide
CAS 82374-34-3
Molecular Formula C16H34OS
Formula Weight 274.51g/mol
MDL Number MFCD00040086
Beilstein 4973978