missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(2-ethylhexyl) sebacate, 97%, ACROS Organics™

$111.89 - $812.32

Chemical Identifiers

CAS 122-62-3
Molecular Formula C26H50O4
Molecular Weight (g/mol) 426.67
InChI Key VJHINFRRDQUWOJ-UHFFFAOYSA-N
Synonym bis 2-ethylhexyl decanedioate
PubChem CID 31218
IUPAC Name bis(2-ethylhexyl) decanedioate
SMILES CCCCC(CC)COC(=O)CCCCCCCCC(=O)OCC(CC)CCCC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC269920010 SDS View Documents Fisher Scientific Acros
269920010
1L Glass bottle Each for $111.89
 
AC269920025 SDS View Documents Fisher Scientific Acros
269920025
2.5L Glass bottle Each for $253.20
 
AC269920100 SDS View Documents Thermo Fisher Scientific
AC269920100
10L Plastic drum Each for $812.32
 
Specifications

Chemical Identifiers

122-62-3
426.67
bis 2-ethylhexyl decanedioate
bis(2-ethylhexyl) decanedioate
C26H50O4
VJHINFRRDQUWOJ-UHFFFAOYSA-N
31218
CCCCC(CC)COC(=O)CCCCCCCCC(=O)OCC(CC)CCCC

Specifications

0.15mg KOH/g max.
215°C
Glass bottle
12% (10g, 6 hrs, 200°C) max.
0.91
0.15% Max. (K.F.)
Yellow
1L
96% min. (GC)
02,181
bis 2-ethylhexyl decanedioate
VJHINFRRDQUWOJ-UHFFFAOYSA-N
bis(2-ethylhexyl) decanedioate
31218
Liquid
Bis(2-ethylhexyl) sebacate
0.9100g/mL
Authentic
1.4500 to 1.455
260 to 265mg KOH/g
ca 23 mPa.s (20°C)
212°C (1.0mmHg)
-55°C
122-62-3
C26H50O4
15,1255
Solubility in water: insoluble
CCCCC(CC)COC(=O)CCCCCCCCC(=O)OCC(CC)CCCC
426.67
426.67
97%
Safety and Handling

Safety and Handling

EINECSNumber : 204-558-8