missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bis(2-ethylhexyl) Fumarate 98.0+%, TCI America™
Supplier: TCI America F0117500ML
Specifications
Bis(2-ethylhexyl) Fumarate | |
Colorless | |
C20H36O4 | |
500 mL | |
ROPXFXOUUANXRR-BUHFOSPRSA-N | |
bis(2-ethylhexyl) (E)-but-2-enedioate | |
5370325 | |
≥98.0% (GC) |
141-02-6 | |
185°C | |
MFCD00065144 | |
Fumaric Acid Di(2-ethylhexyl) Ester, Di(2-ethylhexyl) Fumarate, Dioctyl Fumarate, Fumaric Acid Dioctyl Ester | |
CCCCC(CC)COC(=O)C=CC(=O)OCC(CC)CCCC | |
340.504 | |
340.50 | |
Liquid |
Chemical Identifiers
141-02-6 | |
340.504 | |
ROPXFXOUUANXRR-BUHFOSPRSA-N | |
5370325 | |
CCCCC(CC)COC(=O)C=CC(=O)OCC(CC)CCCC |
C20H36O4 | |
MFCD00065144 | |
Fumaric Acid Di(2-ethylhexyl) Ester, Di(2-ethylhexyl) Fumarate, Dioctyl Fumarate, Fumaric Acid Dioctyl Ester | |
bis(2-ethylhexyl) (E)-but-2-enedioate |
Safety and Handling
EINECSNumber : (2)-1099
RTECSNumber : LT0525000
TSCA : Yes