Please login to your online account to display your discounted pricing

Bis(2-diphenylphosphinoethyl)phenylphosphine, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Bis(2-diphenylphosphinoethyl)phenylphosphine
CAS 23582-02-7
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C34H33P3
Formula Weight 534.56
MDL Number MFCD00003048
Synonym TRIPHOS
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC316820050

  SDS
thermo fisher scientific
AC316820050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC316820010

  SDS
thermo fisher scientific
316820010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder and/or Chunks
Formula Weight 534.56
Quantity 5g
Color White to off-white
Melting Point 130° to 134°C
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material Bis(2-diphenylphosphinoethyl)phenylphosphine
Assay Percent Range 97%
Molecular Formula C34H33P3
CAS 23582-02-7
Synonym TRIPHOS
Elemental Analysis 74.1 to 78.7% (C)
Packaging Glass bottle
MDL Number MFCD00003048
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.