missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis[2-(diphenylphosphino)ethyl]ammonium chloride, 97+%, Alfa Aesar™

$47.95 - $527.41

Chemical Identifiers

CAS 66534-97-2
Molecular Formula C28H30ClNP2
Molecular Weight (g/mol) 477.953
MDL Number MFCD00061569
InChI Key AVDUVHMGGDOIQI-UHFFFAOYSA-N
Synonym bis 2-diphenylphosphino ethyl amine hydrochloride, bis 2-diphenylphosphino ethyl ammonium chloride, bis 2-diphenylphosphanyl ethyl amine hydrochloride, bis 2-diphenylphosphino ethyl ammonium chloride, ethanamine, 2-diphenylphosphino-n-2-diphenylphosphino ethyl-, hydrochloride, 2-diphenylphosphanyl-n-2-diphenylphosphanyl ethyl ethan-1-amine-hydrogen chloride 1/1
PubChem CID 21144848
IUPAC Name 2-diphenylphosphanyl-N-(2-diphenylphosphanylethyl)ethanamine;hydrochloride
SMILES C1=CC=C(C=C1)P(CCNCCP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60250MD View Documents Alfa Aesar
H60250MD
250mg Each for $47.95
 
AAH6025003 View Documents Alfa Aesar
H6025003
1g Each for $137.24
 
AAH6025006 View Documents Alfa Aesar
H6025006
5g Each for $527.41
 
Description

Description

Applications
It is involved in the Synthesis of the new chiral aminodiphosphine ligands.

Solubility
Insoluble in water.

Notes
Air Sensitive. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

66534-97-2
477.953
AVDUVHMGGDOIQI-UHFFFAOYSA-N
21144848
C1=CC=C(C=C1)P(CCNCCP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4.Cl
C28H30ClNP2
MFCD00061569
bis 2-diphenylphosphino ethyl amine hydrochloride, bis 2-diphenylphosphino ethyl ammonium chloride, bis 2-diphenylphosphanyl ethyl amine hydrochloride, bis 2-diphenylphosphino ethyl ammonium chloride, ethanamine, 2-diphenylphosphino-n-2-diphenylphosphino ethyl-, hydrochloride, 2-diphenylphosphanyl-n-2-diphenylphosphanyl ethyl ethan-1-amine-hydrogen chloride 1/1
2-diphenylphosphanyl-N-(2-diphenylphosphanylethyl)ethanamine;hydrochloride

Specifications

175°C to 180°C
66534-97-2
[(C6H5)2PCH2CH2]2NH2Cl
bis 2-diphenylphosphino ethyl amine hydrochloride, bis 2-diphenylphosphino ethyl ammonium chloride, bis 2-diphenylphosphanyl ethyl amine hydrochloride, bis 2-diphenylphosphino ethyl ammonium chloride, ethanamine, 2-diphenylphosphino-n-2-diphenylphosphino ethyl-, hydrochloride, 2-diphenylphosphanyl-n-2-diphenylphosphanyl ethyl ethan-1-amine-hydrogen chloride 1/1
C1=CC=C(C=C1)P(CCNCCP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4.Cl
477.953
477.95
Bis[2-(diphenylphosphino)ethyl]ammonium chloride
250mg
C28H30ClNP2
MFCD00061569
AVDUVHMGGDOIQI-UHFFFAOYSA-N
2-diphenylphosphanyl-N-(2-diphenylphosphanylethyl)ethanamine;hydrochloride
21144848
≥97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning