missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis[2-(diphenylphosphino)ethyl]ammonium chloride, 97+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H60250MD

 View more versions of this product

Catalog No. AAH60250MD



Description

Description

It is involved in the Synthesis of the new chiral aminodiphosphine ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is involved in the Synthesis of the new chiral aminodiphosphine ligands.

Solubility
Insoluble in water.

Notes
Air Sensitive. Incompatible with oxidizing agents.
Specifications

Specifications

Bis[2-(diphenylphosphino)ethyl]ammonium chloride
175°C to 180°C
[(C6H5)2PCH2CH2]2NH2Cl
250 mg
bis 2-diphenylphosphino ethyl amine hydrochloride, bis 2-diphenylphosphino ethyl ammonium chloride, bis 2-diphenylphosphanyl ethyl amine hydrochloride, ethanamine, 2-diphenylphosphino-n-2-diphenylphosphino ethyl-, hydrochloride, 2-diphenylphosphanyl-n-2-diphenylphosphanyl ethyl ethan-1-amine-hydrogen chloride 1/1
AVDUVHMGGDOIQI-UHFFFAOYSA-N
2-diphenylphosphanyl-N-(2-diphenylphosphanylethyl)ethanamine;hydrochloride
21144848
≥97%
66534-97-2
C28H30ClNP2
MFCD00061569
Air Sensitive
Insoluble in water.
Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)NCCP(C1=CC=CC=C1)C1=CC=CC=C1
477.95
477.95

Chemical Identifiers

66534-97-2
477.95
AVDUVHMGGDOIQI-UHFFFAOYSA-N
21144848
Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)NCCP(C1=CC=CC=C1)C1=CC=CC=C1
C28H30ClNP2
MFCD00061569
bis 2-diphenylphosphino ethyl amine hydrochloride, bis 2-diphenylphosphino ethyl ammonium chloride, bis 2-diphenylphosphanyl ethyl amine hydrochloride, ethanamine, 2-diphenylphosphino-n-2-diphenylphosphino ethyl-, hydrochloride, 2-diphenylphosphanyl-n-2-diphenylphosphanyl ethyl ethan-1-amine-hydrogen chloride 1/1
2-diphenylphosphanyl-N-(2-diphenylphosphanylethyl)ethanamine;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only