missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(2-[di(1-adamantyl)phosphino]ethyl)amine, 97+%, Alfa Aesar™

$118.12 - $360.85

Chemical Identifiers

CAS 1086138-36-4
Molecular Formula C44H69NP2
Molecular Weight (g/mol) 673.991
MDL Number MFCD17018779
InChI Key KBHQOEPPHGCYHC-UHFFFAOYSA-N
Synonym bis 2-di adamantan-1-yl phosphino ethyl amine, bis 2-di-1-adamantylphosphino ethyl amine, bis 2-bis adamantan-1-yl phosphanyl ethyl amine, bis 2-diadamantylphosphino ethyl amine, bis 2-diadamantylphosphino ethyl amine, bis 2-di 1-adamantyl phosphino ethyl amine, bis 2-di-1-adamantylphosphino ethyl amine,min, bis 2-bis 3r,5s,7s-adamantan-1-yl phosphanyl ethyl amine, 2-di tricyclo 3.3.1.1~3,7~ decan-1-yl phosphanyl-n-2-di tricyclo 3.3.1.1~3,7~ decan-1-yl phosphanyl ethyl ethan-1-amine
PubChem CID 58194772
IUPAC Name 2-[bis(1-adamantyl)phosphanyl]-N-[2-[bis(1-adamantyl)phosphanyl]ethyl]ethanamine
SMILES C1C2CC3CC1CC(C2)(C3)P(CCNCCP(C45CC6CC(C4)CC(C6)C5)C78CC9CC(C7)CC(C9)C8)C12CC3CC(C1)CC(C3)C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60853MD View Documents Alfa Aesar
H60853MD
250mg Each for $118.12
 
AAH6085303 View Documents Alfa Aesar
H6085303
1g Each for $360.85
 
Description

Description

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in refrigerated conditions. Incompatible with oxidizing agents. It is sensitive to air.
Specifications

Chemical Identifiers

1086138-36-4
673.991
KBHQOEPPHGCYHC-UHFFFAOYSA-N
58194772
C1C2CC3CC1CC(C2)(C3)P(CCNCCP(C45CC6CC(C4)CC(C6)C5)C78CC9CC(C7)CC(C9)C8)C12CC3CC(C1)CC(C3)C2
C44H69NP2
MFCD17018779
bis 2-di adamantan-1-yl phosphino ethyl amine, bis 2-di-1-adamantylphosphino ethyl amine, bis 2-bis adamantan-1-yl phosphanyl ethyl amine, bis 2-diadamantylphosphino ethyl amine, bis 2-diadamantylphosphino ethyl amine, bis 2-di 1-adamantyl phosphino ethyl amine, bis 2-di-1-adamantylphosphino ethyl amine,min, bis 2-bis 3r,5s,7s-adamantan-1-yl phosphanyl ethyl amine, 2-di tricyclo 3.3.1.1~3,7~ decan-1-yl phosphanyl-n-2-di tricyclo 3.3.1.1~3,7~ decan-1-yl phosphanyl ethyl ethan-1-amine
2-[bis(1-adamantyl)phosphanyl]-N-[2-[bis(1-adamantyl)phosphanyl]ethyl]ethanamine

Specifications

217°C to 222°C
1086138-36-4
MFCD17018779
KBHQOEPPHGCYHC-UHFFFAOYSA-N
2-[bis(1-adamantyl)phosphanyl]-N-[2-[bis(1-adamantyl)phosphanyl]ethyl]ethanamine
58194772
≥97%
250mg
C44H69NP2
bis 2-di adamantan-1-yl phosphino ethyl amine, bis 2-di-1-adamantylphosphino ethyl amine, bis 2-bis adamantan-1-yl phosphanyl ethyl amine, bis 2-diadamantylphosphino ethyl amine, bis 2-diadamantylphosphino ethyl amine, bis 2-di 1-adamantyl phosphino ethyl amine, bis 2-di-1-adamantylphosphino ethyl amine,min, bis 2-bis 3r,5s,7s-adamantan-1-yl phosphanyl ethyl amine, 2-di tricyclo 3.3.1.1~3,7~ decan-1-yl phosphanyl-n-2-di tricyclo 3.3.1.1~3,7~ decan-1-yl phosphanyl ethyl ethan-1-amine
C1C2CC3CC1CC(C2)(C3)P(CCNCCP(C45CC6CC(C4)CC(C6)C5)C78CC9CC(C7)CC(C9)C8)C12CC3CC(C1)CC(C3)C2
673.991
673.97
Bis(2-[di(1-adamantyl)phosphino]ethyl)amine