Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(2-cyanoethyl)phenylphosphine

Chemical Identifiers

Chemical Name or Material Bis(2-cyanoethyl)phenylphosphine
CAS 15909-92-9
Formula Weight 216.22
MDL Number MFCD00013827
UN Number UN3464
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3926803

  SDS
alfa aesar™
3926803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3926806

  SDS
alfa aesar™
3926806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 216.22
MDL Number MFCD00013827
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 72° to 73°C
Health Hazard 1 Warning
Chemical Name or Material Bis(2-cyanoethyl)phenylphosphine
Physical Form Crystalline
UN Number UN3464
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, TOXIC, SOLID, N.O.S.
Boiling Point 176° to 178°C (0.5mmHg)
Quantity 1g
CAS 15909-92-9
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.