Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(2-cyanoethyl)amine, tech. 90%

$45.81 - $473.12

Chemical Identifiers

Chemical Name or Material Bis(2-cyanoethyl)amine
CAS 111-94-4
Purity Grade Technical Grade
Molecular Formula HN(CH2CH2CN)2
Formula Weight 123.16
MDL Number MFCD00001956
Synonym Iminobis(propionitrile); Iminodi(propionitrile)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA199590E

  SDS
alfa aesar™
A199590E
2500g Each for $473.12

AAA1995922

  SDS
alfa aesar™
A1995922
100g Each for $45.81

AAA1995936

  SDS
alfa aesar™
A1995936
500g Each for $106.84
Description & Specifications

Specifications

Formula Weight 123.16
Chemical Name or Material Bis(2-cyanoethyl)amine
TSCA TSCA
Purity Grade Technical Grade
Assay 90%
Molecular Formula HN(CH2CH2CN)2
MDL Number MFCD00001956
CAS 111-94-4
Boiling Point 204° to 206°C (25mmHg)
Synonym Iminobis(propionitrile); Iminodi(propionitrile)
Quantity 2500g
Flash Point >110°C (230°F)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
RTECS Number UG2975000

Applications
Bis(2-cyanoethyl)amine is used as pharmaceutical intermediates.

Solubility
Soluble in water.

Notes
Store at 4°C. Store away from moisture and oxidizing agents under dry inert gas. Incompatible with moisture and oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

2° to 8°C