Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(2-chlorophenyl) phosphorochloridate, 97%

Chemical Identifiers

Chemical Name or Material Bis(2-chlorophenyl) phosphorochloridate
CAS 17776-78-2
Molecular Formula C12H8Cl3O3P
Formula Weight 337.53
MDL Number MFCD00015736
UN Number UN3265
Beilstein 2152478
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0263006

  SDS
alfa aesar™
L0263006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0263014

  SDS
alfa aesar™
L0263014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 337.53
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Chemical Name or Material Bis(2-chlorophenyl) phosphorochloridate
Molecular Formula C12H8Cl3O3P
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00015736
Assay 97%
CAS 17776-78-2
Boiling Point 184° to 186°C (0.4mmHg)
Health Hazard 1 Danger
Quantity 5g
Beilstein 2152478
UN Number UN3265
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.