Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(2-bromobenzyl)amine hydrochloride, 99%

$128.82 - $1,660.45

Chemical Identifiers

Chemical Name or Material Bis(2-bromobenzyl)amine hydrochloride
CAS 336615-45-3
Molecular Formula C14H13Br2N·HCl
Formula Weight 391.53
MDL Number MFCD01632133
Synonym 2,2'-Dibromodibenzylamine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2131806

  SDS
alfa aesar™
B2131806
5g Each for $128.82

AAB2131814

  SDS
alfa aesar™
B2131814
25g Each for $495.73

AAB2131822

  SDS
alfa aesar™
B2131822
100g Each for $1,660.45
Description & Specifications

Specifications

Formula Weight 391.53
Synonym 2,2'-Dibromodibenzylamine hydrochloride
Molecular Formula C14H13Br2N·HCl
CAS 336615-45-3
Health Hazard 1 Warning
Chemical Name or Material Bis(2-bromobenzyl)amine hydrochloride
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 99%
MDL Number MFCD01632133
Melting Point 165° to 168°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.