missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bis(2-bromobenzyl)amine hydrochloride, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2131822
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Bis(2-bromobenzyl)amine hydrochloride | |
165°C to 168°C | |
MFCD01632133 | |
bis 2-bromobenzyl amine hydrochloride, bis 2-bromophenyl methyl amine hydrochloride, acmc-20aloa, bis 2-bromobenzyl ;amine hydrochloride, 2,2'-dibromodibenzylamine hydrochloride, bis-2-bromobenzyl amine hydrochloride, 1-2-bromophenyl-n-2-bromophenyl methyl methanamine-hydrogen chloride 1/1 | |
C1=CC=C(C(=C1)CNCC2=CC=CC=C2Br)Br.Cl | |
391.531 | |
391.53 |
336615-45-3 | |
C14H14Br2ClN | |
100 g | |
KXMULHDKALNZDJ-UHFFFAOYSA-N | |
1-(2-bromophenyl)-N-[(2-bromophenyl)methyl]methanamine;hydrochloride | |
24212083 | |
99% |
Chemical Identifiers
336615-45-3 | |
391.531 | |
KXMULHDKALNZDJ-UHFFFAOYSA-N | |
24212083 | |
C1=CC=C(C(=C1)CNCC2=CC=CC=C2Br)Br.Cl |
C14H14Br2ClN | |
MFCD01632133 | |
bis 2-bromobenzyl amine hydrochloride, bis 2-bromophenyl methyl amine hydrochloride, acmc-20aloa, bis 2-bromobenzyl ;amine hydrochloride, 2,2'-dibromodibenzylamine hydrochloride, bis-2-bromobenzyl amine hydrochloride, 1-2-bromophenyl-n-2-bromophenyl methyl methanamine-hydrogen chloride 1/1 | |
1-(2-bromophenyl)-N-[(2-bromophenyl)methyl]methanamine;hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only