missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(2-benzamidophenyl) disulfide, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1211022

 View more versions of this product

Catalog No. AAL1211022



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Bis(2-benzamidophenyl) disulfide
140°C to 143°C
MFCD00043806
721443
ZHMIOPLMFZVSHY-UHFFFAOYSA-N
N-[2-[(2-benzamidophenyl)disulfanyl]phenyl]benzamide
67271
97%
135-57-9
C26H20N2O2S2
100 g
bis 2-benzamidophenyl disulfide, peptazin bafd, peptisant 1o, pepton 22, 2,2'-dibenzoylaminodiphenyl disulfide, bis o-benzamidophenyl disulfide, o,o'-dibenzamidodiphenyl disulfide, 2,2'-dibenzamidodiphenyl disulfide, n,n'-dithiodi-o-phenylenedibenzamide, 2',2'-dithiodibenzanilide
C1=CC=C(C=C1)C(=O)NC2=CC=CC=C2SSC3=CC=CC=C3NC(=O)C4=CC=CC=C4
456.578
456.59

Chemical Identifiers

135-57-9
456.578
ZHMIOPLMFZVSHY-UHFFFAOYSA-N
67271
C1=CC=C(C=C1)C(=O)NC2=CC=CC=C2SSC3=CC=CC=C3NC(=O)C4=CC=CC=C4
C26H20N2O2S2
MFCD00043806
bis 2-benzamidophenyl disulfide, peptazin bafd, peptisant 1o, pepton 22, 2,2'-dibenzoylaminodiphenyl disulfide, bis o-benzamidophenyl disulfide, o,o'-dibenzamidodiphenyl disulfide, 2,2'-dibenzamidodiphenyl disulfide, n,n'-dithiodi-o-phenylenedibenzamide, 2',2'-dithiodibenzanilide
N-[2-[(2-benzamidophenyl)disulfanyl]phenyl]benzamide
Safety and Handling

Safety and Handling

EINECSNumber : 205-201-9

RTECSNumber : CV8700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only