missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(2,4-pentanedionato)molybdenum(VI) Dioxide 95.0+%, TCI America™

Manufacturer:  TCI America M04645G

 View more versions of this product

Catalog No. M04645G



Specifications

Specifications

Bis(2,4-pentanedionato)molybdenum(VI) Dioxide
5g
C10H14MoO6
Acetylacetone Molybdenum(VI)dioxy Salt, Bis(acetylacetonato)dioxomolybdenum(VI), Molybdenum(VI)dioxy Acetylacetonate
Crystal-Powder at 20°C
Undesignated
17524-05-9
MFCD00011506
326.18
≥95.0% (T)

Chemical Identifiers

17524-05-9
C10H14MoO6
MFCD00011506
Acetylacetone Molybdenum(VI)dioxy Salt, Bis(acetylacetonato)dioxomolybdenum(VI), Molybdenum(VI)dioxy Acetylacetonate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2181

TSCA : No

Documents