missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(2,4-pentanedionato)manganese(II) Dihydrate, TCI America™

Manufacturer:  TCI America M004225G

 View more versions of this product

Catalog No. M004225G



Specifications

Specifications

Bis(2,4-pentanedionato)manganese(II) Dihydrate
25g
C10H20MnO6
bis 4-hydroxypent-3-en-2-one dihydrate manganese
CC(=CC(=O)C)O.CC(=CC(=O)C)O.O.O.[Mn]
291.202
253.16
≥97.0% (T)
Yellow
14024-58-9
MFCD00000022
XBBNRHYLXGCQDT-VGKOASNMSA-N
(Z)-4-hydroxypent-3-en-2-one;manganese;dihydrate
54669727
Crystal-Powder at 20°C

Chemical Identifiers

14024-58-9
291.202
XBBNRHYLXGCQDT-VGKOASNMSA-N
54669727
CC(=CC(=O)C)O.CC(=CC(=O)C)O.O.O.[Mn]
C10H20MnO6
MFCD00000022
bis 4-hydroxypent-3-en-2-one dihydrate manganese
(Z)-4-hydroxypent-3-en-2-one;manganese;dihydrate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2142

RTECSNumber : OO9350000

TSCA : Yes

Documents