missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(2,4-pentanedionato)magnesium(II), TCI America™

Manufacturer:  TCI America M000125G

 View more versions of this product

Catalog No. M000125G



Specifications

Specifications

Bis(2,4-pentanedionato)magnesium(II)
276°C
14024-56-7
MFCD00013497
AKTIAGQCYPCKFX-SYWGCQIGSA-L
magnesium;(E)-4-oxopent-2-en-2-olate
25113476
Crystal-Powder at 20°C
White
25g
C10H14MgO4
4-4-oxopent-2-en-2-yl oxy magnesio oxy pent-3-en-2-one
CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].[Mg+2]
222.523
222.52
≥98.0% (T)

Chemical Identifiers

14024-56-7
222.523
AKTIAGQCYPCKFX-SYWGCQIGSA-L
25113476
CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].[Mg+2]
C10H14MgO4
MFCD00013497
4-4-oxopent-2-en-2-yl oxy magnesio oxy pent-3-en-2-one
magnesium;(E)-4-oxopent-2-en-2-olate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2181

TSCA : No

Documents