Please login to your online account to display your discounted pricing

Bis(2,3,4,5-tetramethyl-2,4-cyclopentadiene-1-yl)dimethylsilane, +95%, ACROS Organics™


Quantity 100mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC388901000

 View more versions of this product

Catalog No. AC388901000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥95.0%
Physical Form Powder and Chunks
Formula Weight 300.56
Quantity 100mg
Color Yellow
CAS Min % 95.0
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Bis(2,3,4,5-tetramethyl-2,4-cyclopentadiene-1-yl)dimethylsilane
Molecular Formula C20H32Si
MDL Number MFCD00674017
CAS 89597-05-7
Assay Percent Range 95+%

Specifications

Chemical Name or Material Bis(2,3,4,5-tetramethyl-2,4-cyclopentadiene-1-yl)dimethylsilane
CAS 89597-05-7
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95.0%
Assay Percent Range 95+%
Molecular Formula C20H32Si
Formula Weight 300.56
MDL Number MFCD00674017