Please login to your online account to display your discounted pricing

Bis(2,3,4,5-tetramethyl-2,4-cyclopentadiene-1-yl)dimethylsilane, +95%, ACROS Organics™

$450.11 - $450.11

Chemical Identifiers

Chemical Name or Material Bis(2,3,4,5-tetramethyl-2,4-cyclopentadiene-1-yl)dimethylsilane
CAS 89597-05-7
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95.0%
Assay Percent Range 95+%
Molecular Formula C20H32Si
Formula Weight 300.56
MDL Number MFCD00674017
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC388901000

  SDS
thermo fisher scientific
AC388901000
Glass bottle 100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC388905000

  SDS
thermo fisher scientific
AC388905000
Glass bottle 500mg Each for $450.11
Description & Specifications

Specifications

Assay ≥95.0%
Physical Form Powder and Chunks
Formula Weight 300.56
Quantity 100mg
Color Yellow
CAS Min % 95.0
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Bis(2,3,4,5-tetramethyl-2,4-cyclopentadiene-1-yl)dimethylsilane
Molecular Formula C20H32Si
MDL Number MFCD00674017
CAS 89597-05-7
Assay Percent Range 95+%