missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(2,2-diethoxyethyl) Disulfide, TCI America™

$161.95 - $546.95

Chemical Identifiers

CAS 76505-71-0
Molecular Formula C12H26O4S2
Molecular Weight (g/mol) 298.456
InChI Key XJIKEAQNTPCOLU-UHFFFAOYSA-N
PubChem CID 10990187
IUPAC Name 2-(2,2-diethoxyethyldisulfanyl)-1,1-diethoxyethane
SMILES CCOC(CSSCC(OCC)OCC)OCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B28061G View Documents TCI America
B28061G
1g Each for $161.95
 
B28065G View Documents TCI America
B28065G
5g Each for $546.95
 
Specifications

Chemical Identifiers

76505-71-0
298.456
10990187
CCOC(CSSCC(OCC)OCC)OCC
C12H26O4S2
XJIKEAQNTPCOLU-UHFFFAOYSA-N
2-(2,2-diethoxyethyldisulfanyl)-1,1-diethoxyethane

Specifications

155°C
1g
C12H26O4S2
CCOC(CSSCC(OCC)OCC)OCC
298.456
298.46
≥90.0% (GC)
Undesignated
76505-71-0
XJIKEAQNTPCOLU-UHFFFAOYSA-N
2-(2,2-diethoxyethyldisulfanyl)-1,1-diethoxyethane
10990187
Clear Liquid at 20°C
Bis(2,2-diethoxyethyl) Disulfide
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer