missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bis[2-(2-benzothiazolyl)phenolato]zinc(II) 98.0+%, TCI America™
$54.75 - $164.24
Chemical Identifiers
CAS | 58280-31-2 |
---|---|
Molecular Formula | C26H18N2O2S2Zn |
Molecular Weight (g/mol) | 519.942 |
MDL Number | MFCD03844766 |
InChI Key | MPDFTJIWQKPYAF-YJCYQSLBSA-N |
PubChem CID | 50878549 |
IUPAC Name | (6E)-6-(3H-1,3-benzothiazol-2-ylidene)cyclohexa-2,4-dien-1-one;zinc |
SMILES | C1=CC=C2C(=C1)NC(=C3C=CC=CC3=O)S2.C1=CC=C2C(=C1)NC(=C3C=CC=CC3=O)S2.[Zn] |
Chemical Identifiers
58280-31-2 | |
519.942 | |
MPDFTJIWQKPYAF-YJCYQSLBSA-N | |
(6E)-6-(3H-1,3-benzothiazol-2-ylidene)cyclohexa-2,4-dien-1-one;zinc |
C26H18N2O2S2Zn | |
MFCD03844766 | |
50878549 | |
C1=CC=C2C(=C1)NC(=C3C=CC=CC3=O)S2.C1=CC=C2C(=C1)NC(=C3C=CC=CC3=O)S2.[Zn] |
Specifications
58280-31-2 | |
MFCD03844766 | |
MPDFTJIWQKPYAF-YJCYQSLBSA-N | |
(6E)-6-(3H-1,3-benzothiazol-2-ylidene)cyclohexa-2,4-dien-1-one;zinc | |
50878549 | |
≥98.0% (T) | |
Bis[2-(2-benzothiazolyl)phenolato]zinc(II) |
C26H18N2O2S2Zn | |
1 g | |
C1=CC=C2C(=C1)NC(=C3C=CC=CC3=O)S2.C1=CC=C2C(=C1)NC(=C3C=CC=CC3=O)S2.[Zn] | |
519.942 | |
517.93 | |
Crystalline Powder |
Safety and Handling
TSCA : No