Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)magnesium(II) hydrate, 98+%

$96.28 - $96.28

Chemical Identifiers

Chemical Name or Material Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)magnesium(II) hydrate
CAS 21361-35-3
Formula Weight 390.85 (Anhydrous)
MDL Number MFCD00167208
Synonym Magnesium-DPM dihydrate; Mg(TMHD)2
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3859903

  SDS
alfa aesar™
3859903
1g Each for $96.28

AA3859906

  SDS
alfa aesar™
3859906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 390.85 (Anhydrous)
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00167208
Physical Form Powder
Quantity 1g
Synonym Magnesium-DPM dihydrate; Mg(TMHD)2
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 135° to 150°C (° to H2O)
Boiling Point 150°C (0.05mmHg) (subl.)
CAS 21361-35-3
Health Hazard 1 Warning
Chemical Name or Material Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)magnesium(II) hydrate
Assay ≥98%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.