Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)lead(II)

Chemical Identifiers

Chemical Name or Material Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)lead(II)
CAS 21319-43-7
Formula Weight 573.75
MDL Number MFCD00070468
UN Number UN3467
Synonym Lead(II)-DPM; Pb(TMHD)2
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3939703

  SDS
alfa aesar™
3939703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3939706

  SDS
alfa aesar™
3939706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 573.75
Melting Point 126° to 128°C
Boiling Point 134°C (0.1mmHg) (subl.)
UN Number UN3467
Physical Form Powder
CAS 21319-43-7
MDL Number MFCD00070468
Health Hazard 2 GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Quantity 1g
Health Hazard 3 GHS P Statement
P260-P261-P281-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOMETALLIC COMPOUND, SOLID, TOXIC, N.O.S.
Synonym Lead(II)-DPM; Pb(TMHD)2
Health Hazard 1 Danger
Chemical Name or Material Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)lead(II)
Safety and Handling

Danger

GHS P Statement
P260-P261-P281-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.