Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)copper(II), 99.9% (metals basis)

$204.92 - $890.50

Chemical Identifiers

Chemical Name or Material Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)copper(II)
CAS 14040-05-2
Formula Weight 430.05
MDL Number MFCD00058920
Synonym Copper(II)-DPM; Cu(TMHD)2
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4127906

  SDS
alfa aesar™
4127906
5g Each for $204.92

AA4127914

  SDS
alfa aesar™
4127914
25g Each for $890.50

AA4127903

  SDS
alfa aesar™
4127903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)copper(II)
Boiling Point 100°C (0.1mmHg) (subl.)
Melting Point 198°C
CAS 14040-05-2
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Physical Form Powder
Assay 99.9%
Assay Percent Notes (metals basis)
Formula Weight 430.05
MDL Number MFCD00058920
Synonym Copper(II)-DPM; Cu(TMHD)2
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.