Learn More
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(II), 99.9% (metals basis), Thermo Scientific Chemicals
Precursor for CVD process
$118.38 - $1384.04
Chemical Identifiers
CAS | 13986-53-3 |
---|---|
Molecular Formula | C22H38CoO4 |
Molecular Weight (g/mol) | 425.48 |
MDL Number | MFCD00233616 |
InChI Key | KLJJOSZRALJWDS-UHFFFAOYSA-N |
Synonym | bis 2,2,6,6-tetramethyl-3,5-heptanedionato cobalt ii, cobalt 2+ bis 2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate |
PubChem CID | 57369563 |
IUPAC Name | cobalt(2+);2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate |
SMILES | [Co++].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA4135902
|
Thermo Scientific Chemicals
04135902 |
0.5 g |
Each for $118.38
|
|
|||||
AA4135904
|
Thermo Scientific Chemicals
04135904 |
2 g |
Each for $363.00
|
|
|||||
AA4135909
|
Thermo Scientific Chemicals
04135909 |
10 g |
Each for $1,384.04
|
|
|||||
Description
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(II) is used in nano-material, thin films and ferroelectric metals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13986-53-3 | |
425.48 | |
KLJJOSZRALJWDS-UHFFFAOYSA-N | |
57369563 | |
[Co++].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C |
C22H38CoO4 | |
MFCD00233616 | |
bis 2,2,6,6-tetramethyl-3,5-heptanedionato cobalt ii, cobalt 2+ bis 2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate | |
cobalt(2+);2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate |
Specifications
13986-53-3 | |
C22H38CoO4 | |
0.5 g | |
Soluble in hydrocarbons,alcohols and ethers. Slightly soluble in water. | |
[Co++].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C | |
425.48 | |
425.46 | |
Powder | |
(metals basis) |
143°C | |
MFCD00233616 | |
bis 2,2,6,6-tetramethyl-3,5-heptanedionato cobalt ii, cobalt 2+ bis 2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate | |
KLJJOSZRALJWDS-UHFFFAOYSA-N | |
cobalt(2+);2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate | |
57369563 | |
99.9% | |
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(II) |
Safety and Handling
GHS H Statement
H341-H317
Suspected of causing genetic defects.
May cause an allergic skin reaction.
P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H341
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only