missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)barium(II) hydrate, 98+%

MOCVD of superconductors

Manufacturer:  Alfa Aesar™ 1453903

 View more versions of this product

Catalog No. AA1453903



Specifications

Specifications

Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)barium(II) hydrate
225°C (0.3mmHg)
172°C
17594-47-7
MFCD00058709
barium-dpm, ba tmhd 2, bis 2,2,6,6-tetramethyl-3,5-heptanedionato barium ii, barium 2,2,6,6-tetramethyl-3,5-heptanedionate tetramer, barium bis 2,2,6,6-tetramethyl-3,5-heptanedionate hydrate, bis 2,2,6,6-tetramethyl-3,5-heptanedionate barium ii, barium-thd, barium bis dipivaloylmethanate, barium ii 2,2,6,6-tetramethylheptanedionate, bis 2,2,6,6-tetramethyl-3,5 heptanedionato barium ii
CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].O.[Ba+2]
521.884
503.85 (Anhydrous)
≥98%
Hygroscopic
White
1g
C22H40BaO5
UN1564
VCALGUJWYYNHDY-ZJCTYWPYSA-L
barium(2+);(Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate;hydrate
17749137
Powder

Chemical Identifiers

17594-47-7
521.884
VCALGUJWYYNHDY-ZJCTYWPYSA-L
17749137
CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].CC(C)(C)C(=CC(=O)C(C)(C)C)[O-].O.[Ba+2]
C22H40BaO5
MFCD00058709
barium-dpm, ba tmhd 2, bis 2,2,6,6-tetramethyl-3,5-heptanedionato barium ii, barium 2,2,6,6-tetramethyl-3,5-heptanedionate tetramer, barium bis 2,2,6,6-tetramethyl-3,5-heptanedionate hydrate, bis 2,2,6,6-tetramethyl-3,5-heptanedionate barium ii, barium-thd, barium bis dipivaloylmethanate, barium ii 2,2,6,6-tetramethylheptanedionate, bis 2,2,6,6-tetramethyl-3,5 heptanedionato barium ii
barium(2+);(Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.

Documents