Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)barium(II), 99.99% (metals basis excluding Sr) Sr< 500ppm

Chemical Identifiers

Chemical Name or Material Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)barium(II)
CAS 17594-47-7
Formula Weight 503.85
MDL Number MFCD00058709
UN Number UN1564
Synonym Barium-DPM
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4134003

  SDS
alfa aesar™
4134003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4134006

  SDS
alfa aesar™
4134006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 503.85
Chemical Name or Material Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)barium(II)
Physical Form Powder
Assay 99.99%
Assay Percent Notes (metals basis excluding Sr) Sr < 500ppm
MDL Number MFCD00058709
CAS 17594-47-7
UN Number UN1564
Synonym Barium-DPM
Melting Point 200°C
Boiling Point 225°C (0.3mmHg)
Quantity 1g
Health Hazard 1 Warning
Sensitivity Hygroscopic
Health Hazard 2 GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
Safety and Handling

Warning

GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.