missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(2,2,2-trifluoroethyl) Ether, TCI America™

$80.97 - $230.70

Chemical Identifiers

CAS 333-36-8
Molecular Formula C4H4F6O
Molecular Weight (g/mol) 182.065
MDL Number MFCD00010604
InChI Key KGPPDNUWZNWPSI-UHFFFAOYSA-N
Synonym 2,2,2-Trifluoroethyl Ether
PubChem CID 9528
IUPAC Name 1,1,1-trifluoro-2-(2,2,2-trifluoroethoxy)ethane
SMILES C(C(F)(F)F)OCC(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B12931G View Documents TCI America
B12931G
1g Each for $80.97
 
B12935G View Documents TCI America
B12935G
5g Each for $230.70
 
Specifications

Chemical Identifiers

333-36-8
182.065
KGPPDNUWZNWPSI-UHFFFAOYSA-N
9528
C(C(F)(F)F)OCC(F)(F)F
C4H4F6O
MFCD00010604
2,2,2-Trifluoroethyl Ether
1,1,1-trifluoro-2-(2,2,2-trifluoroethoxy)ethane

Specifications

64°C
1g
C4H4F6O
3271
KGPPDNUWZNWPSI-UHFFFAOYSA-N
1,1,1-trifluoro-2-(2,2,2-trifluoroethoxy)ethane
9528
Clear Liquid at 20°C
Bis(2,2,2-trifluoroethyl) Ether
Colorless
333-36-8
MFCD00010604
2,2,2-Trifluoroethyl Ether
C(C(F)(F)F)OCC(F)(F)F
182.065
182.07
≥99.0% (GC)
Safety and Handling

Safety and Handling

RTECSNumber : KN3675000

TSCA : No