Please login to your online account to display your discounted pricing

Bis(1H-inden-1-yl)dimethylsilane 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC388480010

 View more versions of this product

Catalog No. AC388480010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 288.46
Quantity 1g
Color Yellow to light brown
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material Bis(1H-inden-1-yl)dimethylsilane
Packaging Glass bottle
Assay Percent Range 97%
MDL Number MFCD00674015
CAS 18666-26-7
Infrared Spectrum Authentic
Molecular Formula C20H20Si

Specifications

Chemical Name or Material Bis(1H-inden-1-yl)dimethylsilane
CAS 18666-26-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C20H20Si
Formula Weight 288.46
MDL Number MFCD00674015