Please login to your online account to display your discounted pricing

Bis(1H-inden-1-yl)dimethylsilane 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Bis(1H-inden-1-yl)dimethylsilane
CAS 18666-26-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C20H20Si
Formula Weight 288.46
MDL Number MFCD00674015
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC388480010

  SDS
thermo fisher scientific
AC388480010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC388480050

  SDS
thermo fisher scientific
AC388480050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 288.46
Quantity 1g
Color Yellow to light brown
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material Bis(1H-inden-1-yl)dimethylsilane
Packaging Glass bottle
Assay Percent Range 97%
MDL Number MFCD00674015
CAS 18666-26-7
Infrared Spectrum Authentic
Molecular Formula C20H20Si