Please login to your online account to display your discounted pricing

Bis(1,5-cyclooctadiene)nickel(0), ACROS Organics™


Quantity 25g

Manufacturer: thermo fisher scientific  AC223970250

 View more versions of this product

Catalog No. AC223970250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Shiny Crystalline Powder
Formula Weight 275.06
Color Gold
Quantity 25g
MDL Number MFCD00058902
CAS 1295-35-8
Synonym Nickel(COD)2
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS P Statement Flammable solid. May cause cancer. May cause an allergic skin reaction.
Nickel (Ni) 21% (typically)
Chemical Name or Material Bis(1, 5-cyclooctadiene)nickel(0)
Molecular Formula C16H24Ni
Health Hazard 3 GHS P Statement Do not handle until all safety precautions have been read and understood. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Use personal protective equipment as required. IF exposed or concerned: Get medical advice/attention. Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Obtain special instructions before use.

Specifications

Chemical Name or Material Bis(1, 5-cyclooctadiene)nickel(0)
CAS 1295-35-8
Molecular Formula C16H24Ni
Formula Weight 275.06
MDL Number MFCD00058902
Synonym Nickel(COD)2
Accessories

GHS Signal Word: Danger

GHS P Statement Do not handle until all safety precautions have been read and understood. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Use personal protective equipment as required. IF exposed or concerned: Get medical advice/attention. Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Obtain special instructions before use.

GHS P Statement Flammable solid. May cause cancer. May cause an allergic skin reaction.

SDS & Safety Documents