Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(1,5-cyclooctadiene)nickel(0)

$105.03 - $105.03

Chemical Identifiers

Chemical Name or Material Bis(1,5-cyclooctadiene)nickel(0)
CAS 1295-35-8
Formula Weight 275.06
MDL Number MFCD00058902
UN Number UN1325
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3923402

  SDS
alfa aesar™
3923402
0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3923404

  SDS
alfa aesar™
3923404
2g Each for $105.03

AA3923409

  SDS
alfa aesar™
3923409
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 275.06
Chemical Name or Material Bis(1,5-cyclooctadiene)nickel(0)
Physical Form Crystalline
MDL Number MFCD00058902
CAS 1295-35-8
UN Number UN1325
Color Yellow
Melting Point 60°C (dec.)
Quantity 0.5g
Sensitivity Air & Light Sensitive
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H228-H350-H317
Flammable solid.
May cause cancer.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P210-P261-P280-P363-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
Odor Odorless
RTECS Number QR6135000
Safety and Handling

Danger

GHS P Statement
P210-P261-P280-P363-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H228-H350-H317
Flammable solid.
May cause cancer.
May cause an allergic skin reaction.