Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(1,5-cyclooctadiene)iridium(I) tetrafluoroborate

Chemical Identifiers

Chemical Name or Material Bis(1,5-cyclooctadiene)iridium(I) tetrafluoroborate
CAS 35138-23-9
Formula Weight 495.4
MDL Number MFCD00079664
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3941403

  SDS
alfa aesar™
3941403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA39414ME

  SDS
alfa aesar™
39414ME
500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3941499

  SDS
alfa aesar™
3941499
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 495.4
Physical Form Powder
Chemical Name or Material Bis(1,5-cyclooctadiene)iridium(I) tetrafluoroborate
MDL Number MFCD00079664
CAS 35138-23-9
Quantity 1g
Solubility Information Soluble in chloroform, methylene chloride