missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis[1,2-bis(diphenylphosphino)ethane]palladium(0), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 03923302

 View more versions of this product

Catalog No. AA3923302



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Bis[1,2-bis(diphenylphosphino)ethane]palladium(0)
C52H48P4Pd
0.5 g
bis 1,2-bis diphenylphosphino ethane palladium 0, pd dppe 2, pd diphos 2, bis diphos palladium 0, bis 1,2-bis diphenylphosphino ethane palladium, bis diphos palladium, bis 1,2-bis diphenylphosphino ethane-palladium 0, bis 1,2-bis diphenylphosphino ethane-palladium
[Pd].C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
903.27
903.25
31277-98-2
MFCD00009880
Air Sensitive
FAFGMAGIYHHRKN-UHFFFAOYSA-N
2-diphenylphosphanylethyl(diphenyl)phosphane;palladium
2724231
Powder

Chemical Identifiers

31277-98-2
903.27
FAFGMAGIYHHRKN-UHFFFAOYSA-N
2724231
[Pd].C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
C52H48P4Pd
MFCD00009880
bis 1,2-bis diphenylphosphino ethane palladium 0, pd dppe 2, pd diphos 2, bis diphos palladium 0, bis 1,2-bis diphenylphosphino ethane palladium, bis diphos palladium, bis 1,2-bis diphenylphosphino ethane-palladium 0, bis 1,2-bis diphenylphosphino ethane-palladium
2-diphenylphosphanylethyl(diphenyl)phosphane;palladium
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only