missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Biphenylene, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC279740500

 View more versions of this product

Catalog No. AC279740500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Biphenylene
Glass bottle
109°C to 112°C
99+%
99.0
99+%
MFCD00001110
1,1'-biphenylene, 8ci 9ci, biphenylen, biphenylene, biphenylene, 99+%, cyclobutadibenzene, dibenzocyclobutadiene, diphenylene, pubchem9106, unii-0z64i7d5m2
IFVTZJHWGZSXFD-UHFFFAOYSA-N
biphenylene
9214
152.2
99+%
Authentic
Yellow
50mg
259-79-0
100.0
C12H8
05, 298
Solubility in water: 9.60mg/l (calc). Other solubilities: organic solvents general
C1=CC=C2C(=C1)C3=CC=CC=C23
152.2
CHEBI:33079
Needles

Chemical Identifiers

259-79-0
152.2
IFVTZJHWGZSXFD-UHFFFAOYSA-N
9214
biphenylene
C12H8
MFCD00001110
1,1'-biphenylene, 8ci 9ci, biphenylen, biphenylene, biphenylene, 99+%, cyclobutadibenzene, dibenzocyclobutadiene, diphenylene, pubchem9106, unii-0z64i7d5m2
CHEBI:33079
C1=CC=C2C(=C1)C3=CC=CC=C23
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic to aquatic life with long lasting effects.

GHS P Statement:
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents