Learn More
Biphenyl, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 106250010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Biphenyl | |
98.5 | |
68.0°C to 72.0°C | |
255.0°C | |
Authentic | |
Plastic bottle | |
C6H5C6H5 | |
1 kg | |
15, 3344 | |
Solubility in water: insoluble. | |
C1=CC=C(C=C1)C1=CC=CC=C1 | |
154.21 | |
CHEBI:17097 | |
99% |
92-52-4 | |
100.0 | |
White to Yellow | |
113°C | |
98.5% min. (GC) | |
C12H10 | |
MFCD00003054 | |
05, 578 | |
biphenyl, phenylbenzene, diphenyl, 1,1'-diphenyl, bibenzene, lemonene, xenene, phenador-x, tetrosin ly, carolid al | |
ZUOUZKKEUPVFJK-UHFFFAOYSA-N | |
1,1'-biphenyl | |
7095 | |
154.21 | |
Crystalline Powder, Crystals or Flakes |
Chemical Identifiers
92-52-4 | |
154.21 | |
ZUOUZKKEUPVFJK-UHFFFAOYSA-N | |
7095 | |
1,1'-biphenyl |
C12H10 | |
MFCD00003054 | |
biphenyl, phenylbenzene, diphenyl, 1,1'-diphenyl, bibenzene, lemonene, xenene, phenador-x, tetrosin ly, carolid al | |
CHEBI:17097 | |
C1=CC=C(C=C1)C1=CC=CC=C1 |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/att
GHS Signal Word: Warning
EINECSNumber : 202-163-5
RUO – Research Use Only