missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Biphenyl, 99%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 106252500

 View more versions of this product

Catalog No. AC106252500


 

Specifications Description & Specifications

Specifications

Biphenyl
98.5
Authentic
Crystalline Powder, Crystals or Flakes
99%
C6H5C6H5
MFCD00003054
05, 578
15, 3344
250g
92-52-4
100.0
Plastic bottle
≥98.5%
C12H10
154.21
White to faint yellow
68° to 72°C
Diphenyl

Chemical Identifiers

92-52-4
100.0
C12H10
154.21
Diphenyl
≥98.5%
98.5
99%
C6H5C6H5
MFCD00003054
Biphenyl

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning