Learn More
Biphenyl, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A102650B
Description
Biphenyl used in combination with Li metal for the generation of activated Ca metal. It is also used as important starting material for the production of polychlorinated biphenyls. It is also an intermediate for the production of a host of other organic compounds
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBiphenyl used in combination with Li metal for the generation of activated Ca metal. It is also used as important starting material for the production of polychlorinated biphenyls. It is also an intermediate for the production of a host of other organic compounds
Solubility
Soluble in alcohol and ether
Notes
Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents. keep container tightly closed.
Specifications
Biphenyl | |
69°C to 72°C | |
254°C to 255°C | |
Pleasant | |
MFCD00003054 | |
UN3077 | |
14,3314 | |
Soluble in alcohol and ether | |
C1=CC=C(C=C1)C1=CC=CC=C1 | |
154.21 | |
CHEBI:17097 | |
99% |
92-52-4 | |
0.99 | |
113°C (235°F) | |
C12H10 | |
1000 g | |
1634058 | |
biphenyl, phenylbenzene, diphenyl, 1,1'-diphenyl, bibenzene, lemonene, xenene, phenador-x, tetrosin ly, carolid al | |
ZUOUZKKEUPVFJK-UHFFFAOYSA-N | |
1,1'-biphenyl | |
7095 | |
154.21 |
Chemical Identifiers
92-52-4 | |
154.21 | |
ZUOUZKKEUPVFJK-UHFFFAOYSA-N | |
7095 | |
1,1'-biphenyl |
C12H10 | |
MFCD00003054 | |
biphenyl, phenylbenzene, diphenyl, 1,1'-diphenyl, bibenzene, lemonene, xenene, phenador-x, tetrosin ly, carolid al | |
CHEBI:17097 | |
C1=CC=C(C=C1)C1=CC=CC=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 202-163-5
RTECSNumber : DU8050000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only