missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Biphenyl, 99%

Manufacturer:  Alfa Aesar™ A102650I

 View more versions of this product

Catalog No. AAA102650I



Description

Description

Applications
Biphenyl used in combination with Li metal for the generation of activated Ca metal. It is also used as important starting material for the production of polychlorinated biphenyls. It is also an intermediate for the production of a host of other organic compounds

Solubility
Soluble in alcohol and ether

Notes
Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents. keep container tightly closed.
Specifications

Specifications

Biphenyl
Pleasant
C12H10
UN3077
14,3314
Diphenyl; Phenylbenzene
5000g
Soluble in alcohol and ether
C1=CC=C(C=C1)C2=CC=CC=C2
154.212
CHEBI:17097
92-52-4
99%
MFCD00003054
1634058
69°C to 72°C
254°C to 255°C
113°C (235°F)
ZUOUZKKEUPVFJK-UHFFFAOYSA-N
1,1'-biphenyl
7095
154.21

Chemical Identifiers

92-52-4
154.212
ZUOUZKKEUPVFJK-UHFFFAOYSA-N
7095
1,1'-biphenyl
C12H10
MFCD00003054
Diphenyl; Phenylbenzene
CHEBI:17097
C1=CC=C(C=C1)C2=CC=CC=C2
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : DU8050000

TSCA : TSCA

SDS Documents