Please login to your online account to display your discounted pricing

Alfa Aesar™ Biphenyl-4-carboxamide, 98%

$281.69 - $281.69

Chemical Identifiers

Chemical Name or Material Biphenyl-4-carboxamide
CAS 3815-20-1
Molecular Formula C13H11NO
Formula Weight 197.24
MDL Number MFCD00017573
Beilstein 2614166
Synonym 4-Phenylbenzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1681314

 
alfa aesar™
A1681314
25g Each for $281.69
Description & Specifications

Specifications

Formula Weight 197.24
Chemical Name or Material Biphenyl-4-carboxamide
Assay 98%
Molecular Formula C13H11NO
MDL Number MFCD00017573
CAS 3815-20-1
Beilstein 2614166
Melting Point 227° to 229°C
Synonym 4-Phenylbenzamide
Quantity 25g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Odor Odorless
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.