Please login to your online account to display your discounted pricing

Alfa Aesar™ Biphenyl-4,4'-diboronic acid bis(neopentyl glycol) ester, 98%

$295.78 - $295.78

Chemical Identifiers

Chemical Name or Material Biphenyl-4,4'-diboronic acid bis(neopentyl glycol) ester
CAS 5487-93-4
Molecular Formula C22H28B2O4
Formula Weight 378.09g/mol
MDL Number MFCD01318103
Beilstein 2952067
Synonym Biphenyl-4,4'-diboronic acid bis-2,2-dimethyltrimethylene ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1760506

 
alfa aesar™
L17605-06
5g Each for $295.78
Description & Specifications

Specifications

Odor Odorless
Quantity 5g
Melting Point 242° to 247°C
Quantity 5g
Chemical Name or Material Biphenyl-4,4'-diboronic acid bis(neopentyl glycol) ester
CAS 5487-93-4
Assay 98%
Molecular Formula C22H28B2O4
Formula Weight 378.09g/mol
MDL Number MFCD01318103
Beilstein 2952067
Synonym Biphenyl-4,4'-diboronic acid bis-2,2-dimethyltrimethylene ester