missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Biphenyl-4,4'-diboronic acid, 95%

Manufacturer:  Alfa Aesar™ L1332806

 View more versions of this product

Catalog No. AAL1332806



Description

Description

Applications
Biphenyl-4,4′-diboronic acid is used in the preparation of cycloparaphenylenes through Suzuki coupling. It is also employed in the synthesis of 1,3,2-diazaboroine derivatives, which is useful for organic thin-film transistor applications. It plays an important role in the preparation of arylboronates from reactions with catechol, dihydroxynaphthalene and diaminobenzenedithiol, which finds application as organic field-effect transistors and light emitting diodes.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Biphenyl-4,4'-diboronic acid
5g
C12H12B2O4
2941876
SLHKDOGTVUCXKX-UHFFFAOYSA-N
[4-(4-boronophenyl)phenyl]boronic acid
2734608
95%
~300°C
4151-80-8
MFCD00151795
4,4'-biphenyldiboronic acid, 1,1'-biphenyl-4,4'-diyldiboronic acid, biphenyl-4,4'-diboronic acid, 4-4-boronophenyl phenyl boronic acid, biphenyl-4,4'-diyldiboronic acid, pubchem5324, acmc-1apw8, 4,4'-biphenyldiboric acid, 4,4-biphenyldiboronic acid, 4,4'-diboronobiphenyl
B(C1=CC=C(C=C1)C2=CC=C(C=C2)B(O)O)(O)O
241.844
241.85

Chemical Identifiers

4151-80-8
241.844
SLHKDOGTVUCXKX-UHFFFAOYSA-N
2734608
B(C1=CC=C(C=C1)C2=CC=C(C=C2)B(O)O)(O)O
C12H12B2O4
MFCD00151795
4,4'-biphenyldiboronic acid, 1,1'-biphenyl-4,4'-diyldiboronic acid, biphenyl-4,4'-diboronic acid, 4-4-boronophenyl phenyl boronic acid, biphenyl-4,4'-diyldiboronic acid, pubchem5324, acmc-1apw8, 4,4'-biphenyldiboric acid, 4,4-biphenyldiboronic acid, 4,4'-diboronobiphenyl
[4-(4-boronophenyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents