Please login to your online account to display your discounted pricing

Alfa Aesar™ Biphenyl-3-carboxaldehyde, 95%

$64.65 - $210.95

Chemical Identifiers

Chemical Name or Material Biphenyl-3-carboxaldehyde
CAS 1204-60-0
Molecular Formula C13H10O
Formula Weight 182.22
MDL Number MFCD01740432
Synonym 3-Phenylbenzaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5045303

  SDS
alfa aesar™
H5045303
1g Each for $210.95

AAH50453MD

  SDS
alfa aesar™
H50453MD
250mg Each for $64.65
Description & Specifications

Specifications

Formula Weight 182.22
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C13H10O
Chemical Name or Material Biphenyl-3-carboxaldehyde
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 3-Phenylbenzaldehyde
MDL Number MFCD01740432
Assay 95%
Quantity 1g
RTECS Number DV1771500
CAS 1204-60-0
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.